Learn More
Cole-Parmer Fowler 52-413-006 NIST-Certificate Ruler, 6" L Rigid Steel, 4R Intervals
Catalog No. NC1642206 Shop All Cole-Parmer Products
Encompass_Preferred
Certified rulers are made of hardened and tempered steel and feature easy to read graduations on a non-glare satin chrome finish. Each Fowler Ruler is Pre-Certified for ISO/QS9000 requirements and includes a certificate of calibration.
Catalog No. NC1642206 Supplier Cole-Parmer Supplier No. 0991904
Certified rulers are made of hardened and tempered steel and feature easy to read graduations on a non-glare satin chrome finish. Each Fowler Ruler is Pre-Certified for ISO/QS9000 requirements and includes a certificate of calibration.