Learn More
Cole-Parmer InnoCal NIST-Traceable Calibration, Air, Gas Flowmeter
Supplier:  Cole-Parmer 1708000
Encompass
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s), and test uncertainty ratios (TUR’s) at no additional cost. This NIST-traceable calibration allows you to confirm that the specific asset you purchase meets the manufacturer’s published specification. EPA, USDA, ISO, and GLP/GMPs standards require regular NIST-Traceable calibration of all test equipment being used in critical applications, FDA compliant. Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays.
Catalog No. NC1641193