Learn More
Beckman Coulter NIST Traceable Particle Size Standards
Catalog No. 15655216
Change view
Click to view available options
Particle Size:
500 μm
Material:
Glass
Catalog No. Particle Size Material
15-655-216 500 μm Glass
1 options
This item is not returnable. View return policy
Catalog No. 15-655-216 Supplier Beckman Coulter Supplier No. 7800372/DEL
This item is not returnable. View return policy
Recommended for LS Series Instruments to verify instrument and module performance.
  • For verifying instrument and module performance
  • Recommended for LS Series instruments
  • NIST traceable
TRUSTED_SUSTAINABILITY
Specifications
Product Type Particle Size Standard, GB500
Material Glass
Particle Size 500 μm
Quantity 5 Pk.
For Use With (Application) Verifying Instrument and Module Performance
For Use With (Equipment) LS Series Instruments with ULM & DPS