Learn More
Andwin Scientific NISTmAb, Humanized IgG1k Monoclonal Antibody
SDP
Supplier:  Andwin Scientific RM 8671
Encompass_Preferred
This RM is primarily intended for use in evaluating the performance of methods for determining physiochemical and biophysical attributes of monoclonal antibodies. It also provides a representative test molecule for the development of novel technology in therapeutic protein characterization. Other potential uses include system suitability tests, establishing method or instrument performance and variability, comparing changing analytical test methods, and assisting in method qualification.
Catalog No. NC1554068