Learn More
ATCC NIT-1; Insulinoma; Mouse (Mus musculus)
Catalog No. 502382677
Encompass
NIT-1 is a beta cell line that was isolated from the islet of Langerhans of a 10-week-old, female mouse with insulinoma. This cell line was deposited by EH Leiter.
Catalog No. 50-238-2677 Supplier ATCC Supplier No. CRL2055
Add to Cart
Add to Cart
NIT-1 is a beta cell line that was isolated from the islet of Langerhans of a 10-week-old, female mouse with insulinoma. This cell line was deposited by EH Leiter.