Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nital Etchant 10% Nitric Acid in Ethanol, Reagents

Catalog No. 01334613
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-613 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-613 Supplier Reagents Holdings Llc Supplier No. CN2015001C
 Restricted Item
Add to Cart
Add to Cart

Nital Etchant 10% Nitric Acid in Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 7697-37-2 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Nitric Acid in Ethyl Alcohol
IUPAC Name ethanol
SMILES CCO

Specifications

Color Colorless to Light Yellow
Boiling Point 76°C
Percent Purity 10%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.8703
pH <7
Quantity 1 L
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Nital Etchant 10% Nitric Acid in Ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.