Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nital Etchant, 10% (v/v) Nitric Acid in Reagent Alcohol, Ricca Chemical
SDP

Catalog No. R53230004A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
250 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R53230004A 4 L
R5323000250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R53230004A Supplier Ricca Chemical Company Supplier No. R53230004A
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Nital Etchant
Color Colorless to Yellow
CAS 64-17-5, 7697-37-2, 67-63-0, 67-63-0
CAS Min % 73.0, 16.92, 3.87
CAS Max % 76.0, 17.61, 4.03
Percent Purity 10%
Grade Laboratory
Packaging Natural Poly Bottle
pH <1
Quantity 4 L
Physical Form Liquid
Concentration or Composition Notes 10%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.