Learn More
Nital Etchant 2% Nitric Acid in Ethanol, Reagents
Nital Etchant 2% Nitric Acid in Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$144.30 - $316.00
Chemical Identifiers
| CAS | 64-17-5 , 67-63-0 , 67-56-1 |
|---|---|
| Molecular Formula | C2H6O |
| Molecular Weight (g/mol) | 46.07 |
| InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
| Synonym | Nitric Acid in Ethyl Alcohol |
| IUPAC Name | ethanol |
| SMILES | CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-334-617
|
Reagents Holdings Llc
CN2013001C |
1 L |
Each for $316.00
|
|
|||||
|
01-334-616
|
Reagents Holdings Llc
CN201300500C |
500 mL |
Each for $174.20
|
|
|||||
|
01-334-615
|
Reagents Holdings Llc
CN20130065C |
65 mL |
Each for $144.30
|
|
|||||
Chemical Identifiers
| C2H6O | |
| LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
| ethanol |
Specifications
| Colorless to Light Yellow | |
| 76°C | |
| Nitric Acid in Ethyl Alcohol | |
| CCO | |
| 46.07 | |
| Reagent | |
| 0.8141 | |
| Non-sterile | |
| Nital Etchant 2% Nitric Acid in Ethanol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.