Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nital Etchant 2% Nitric Acid in Ethanol, Reagents

Nital Etchant 2% Nitric Acid in Ethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CN20130065C

Quantity 65 mL

 View more versions of this product

Catalog No. 01-334-615


 Restricted Item
Add to Cart

Description

Description

Specifications

Chemical Identifiers

64-17-5 , 67-63-0 , 67-56-1
46.07
Nitric Acid in Ethyl Alcohol
CCO
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol

Specifications

Colorless to Light Yellow
76°C
Nitric Acid in Ethyl Alcohol
CCO
46.07
Reagent
0.8141
65 mL
Liquid
64-17-5 , 67-63-0 , 67-56-1
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
2%
Glass Bottle
<7
Non-sterile
Nital Etchant 2% Nitric Acid in Ethanol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Accessories
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.