Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nital Etchant, 3% (v/v) in Reagent Alcohol, Ricca Chemical
SDP

Catalog No. R531620025D
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Glass Bottle

Specifications

Chemical Name or Material Nital Etchant, 3% (v/v) in Reagent Alcohol
Color Colorless
CAS 64-17-5, 67-63-0, 67-56-1, 7697-37-2, 7732-18-5
Boiling Point ∼76°C
CAS Min % 85.44, 4.72, 4.27, 3.9, 1.67
CAS Max % 85.44, 4.72, 4.27, 3.9, 1.67
Density 0.81
Solubility Information Miscible in water
Percent Purity 3%
Grade Laboratory
Odor Alcoholic
Packaging Glass Bottle
pH Acidic
Quantity 2.5 L
Flash Point 12.5°C
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.