Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nital Etchant 5% Nitric Acid in Methanol, Reagents

Nital Etchant 5% Nitric Acid in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$70.25 - $182.40

Chemical Identifiers

CAS 67-56-1 , 7697-37-2 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Nitric Acid in Methyl Alcohol
IUPAC Name methanol
SMILES CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-334-648
Accessories View Documents
Reagents Holdings Llc
CN1042001C
1 L
Each for $106.60
Add to Cart
 
 Restricted Item
01-334-647
Accessories View Documents
Reagents Holdings Llc
CN1042002.5D1
2.5 L
Each for $182.40
Add to Cart
 
 Restricted Item
01-334-646
Accessories View Documents
Reagents Holdings Llc
CN104200500C
500 mL
Each for $70.25
Add to Cart
 
 Restricted Item
Description

Description

Specifications

Chemical Identifiers

67-56-1 , 7697-37-2 , 7732-18-5
32.04
Nitric Acid in Methyl Alcohol
CO
CH4O
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol

Specifications

Colorless
64°C
Nitric Acid in Methyl Alcohol
CO
32.04
Reagent
0.8224
Non-sterile
Nital Etchant 5% Nitric Acid in Methanol
67-56-1 , 7697-37-2 , 7732-18-5
CH4O
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
5%
Glass Bottle
<7
Liquid
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.