Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nital Etchant 5% Nitric Acid in Methanol, Reagents

Nital Etchant 5% Nitric Acid in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CN1042002.5D1

Quantity 2.5 L

 View more versions of this product

Catalog No. 01-334-647


Add to Cart

Description

Description

Specifications

Chemical Identifiers

67-56-1 , 7697-37-2 , 7732-18-5
32.04
Nitric Acid in Methyl Alcohol
CO
CH4O
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol

Specifications

Colorless
64°C
Nitric Acid in Methyl Alcohol
CO
32.04
Reagent
0.8224
2.5 L
Liquid
67-56-1 , 7697-37-2 , 7732-18-5
CH4O
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
5%
Glass Bottle
<7
Non-sterile
Nital Etchant 5% Nitric Acid in Methanol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Accessories
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.