Learn More
BTNX Inc Nitazene Test Strip - 100pk (Liquid/Powder)
Catalog No. NC3361238
Encompass_Preferred
The Rapid Response™ Nitazene Test Strip (Liquid/Powder) is a rapid visual immunoassay for the qualitative, presumptive detection of Nitazene in suspicious substances (liquids or powders) at the cut-off concentration of 2000 ng/mL. For forensic use only. Not an IVD.
Catalog No. NC3361238 Supplier BTNX Inc Supplier No. NTZ18S26100
The Rapid Response™ Nitazene Test Strip (Liquid/Powder) is a rapid visual immunoassay for the qualitative, presumptive detection of Nitazene in suspicious substances (liquids or powders) at the cut-off concentration of 2000 ng/mL. For forensic use only. Not an IVD.