Learn More
Cayman Chemical Nitazoxanide, Purity: ≥95%, CAS: 55981-09-4, 50 mg
SDP
Catalog No. 502955669
Encompass_Preferred
A prodrug form of tizoxanide; inhibits C. parvum growth in MDBK cells at 10 µg/ml; active against M. tuberculosis and M. bovis (MIC = 16 µg/ml for both); inhibits SARS-CoV-2 and MERS-CoV replication at 5 and 10 µM; inhibits replication of H1N1 PR8 and WSN, as well as H5N9 A/Ck viruses (EC50s = 1, 0.5, and 1 µg/ml, respectively); reduces oocyst shedding and parasite burden, but not the severity of diarrhea, in a gnotobiotic piglet model of C. parvum infection at 250 mg/kg
Catalog No. 50-295-5669 Supplier Cayman Chemical Supplier No. 1369250 MG
A prodrug form of tizoxanide; inhibits C. parvum growth in MDBK cells at 10 µg/ml; active against M. tuberculosis and M. bovis (MIC = 16 µg/ml for both); inhibits SARS-CoV-2 and MERS-CoV replication at 5 and 10 µM; inhibits replication of H1N1 PR8 and WSN, as well as H5N9 A/Ck viruses (EC50s = 1, 0.5, and 1 µg/ml, respectively); reduces oocyst shedding and parasite burden, but not the severity of diarrhea, in a gnotobiotic piglet model of C. parvum infection at 250 mg/kg