Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nitd-2 | 1197896-79-9 | 99.4% | 433.48 g/mol | C23H19N3O4S | 5 MG
SDP

Catalog No. 5000222696
Encompass_Preferred

NITD-2 is a small-molecule inhibitor of dengue virus (DENV) RNA-dependent RNA polymerase (RdRp) that blocks RdRp-mediated RNA elongation. Supplied for research use, the compound is a solid with limited cell membrane permeability and documented physicochemical and storage properties.

  • Inhibits DENV RdRp-mediated RNA elongation.
  • Small-molecule inhibitor suitable for in vitro antiviral research.
  • Limited cell membrane permeability may affect cellular uptake.
  • High purity: 99.36% (analytical).
  • Molecular formula C23H19N3O4S; molecular weight 433.48 g/mol.
  • Soluble in DMSO (100 mg/mL); formulatable for in vivo use (≥2.5 mg/mL in 10% DMSO/corn oil).
  • Powder stable at -20°C for up to 3 years; solvent stability guidelines available.

Catalog No. 50-002-22696 Supplier Medchemexpress LLC Supplier No. HY1346655MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NITD-2 is a small-molecule inhibitor of dengue virus (DENV) RNA-dependent RNA polymerase (RdRp) that blocks RdRp-mediated RNA elongation. Supplied for research use, the compound is a solid with limited cell membrane permeability and documented physicochemical and storage properties.

  • Inhibits DENV RdRp-mediated RNA elongation.
  • Small-molecule inhibitor suitable for in vitro antiviral research.
  • Limited cell membrane permeability may affect cellular uptake.
  • High purity: 99.36% (analytical).
  • Molecular formula C23H19N3O4S; molecular weight 433.48 g/mol.
  • Soluble in DMSO (100 mg/mL); formulatable for in vivo use (≥2.5 mg/mL in 10% DMSO/corn oil).
  • Powder stable at -20°C for up to 3 years; solvent stability guidelines available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.