Learn More
Medchemexpress LLC NITD-2 | 1197896-79-9 | MFCD34471209 | 99.4% | C23H19N3O4S | 10MG
SDP
Catalog No. 5000261724
Encompass_Preferred
NITD-2 is a small-molecule research inhibitor of dengue virus RNA-dependent RNA polymerase (RdRp). It inhibits RdRp-mediated RNA elongation and is primarily used in biochemical antiviral studies; the compound exhibits limited cellular permeability and is typically prepared in DMSO for assays.
  • Inhibits dengue virus RNA-dependent RNA polymerase (RdRp)-mediated RNA elongation.
  • Limited cell membrane permeability; suitable for in vitro assays and formulated in vivo dosing.
  • High purity appropriate for biochemical and antiviral assays.
  • Soluble in DMSO at high concentrations; ultrasonic agitation may improve solubility.
  • Stable as a powder when stored under recommended cold conditions.
  • Available in small milligram pack sizes for research use.
Catalog No. 50-002-61724 Supplier Medchemexpress LLC Supplier No. HY13466510MG
NITD-2 is a small-molecule research inhibitor of dengue virus RNA-dependent RNA polymerase (RdRp). It inhibits RdRp-mediated RNA elongation and is primarily used in biochemical antiviral studies; the compound exhibits limited cellular permeability and is typically prepared in DMSO for assays.
  • Inhibits dengue virus RNA-dependent RNA polymerase (RdRp)-mediated RNA elongation.
  • Limited cell membrane permeability; suitable for in vitro assays and formulated in vivo dosing.
  • High purity appropriate for biochemical and antiviral assays.
  • Soluble in DMSO at high concentrations; ultrasonic agitation may improve solubility.
  • Stable as a powder when stored under recommended cold conditions.
  • Available in small milligram pack sizes for research use.