Learn More
Medchemexpress LLC HY-109588 5mg Medchemexpress, NITD-349 CAS:1473450-62-2 Purity:>98%
SDP
Catalog No. 501968711
Encompass_Preferred
Medchemexpress, HY-109588 5mg NITD-349 CAS:1473450-62-2 NITD-349 is an MmpL3 inhibitor that shows highly potent anti-mycobacterial activity with MIC50 of 23 nM against virulent Mycobacterium tuberculosis H37Rv. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8711 Supplier Medchemexpress LLC Supplier No. HY1095885MG
Medchemexpress, HY-109588 5mg NITD-349 CAS:1473450-62-2 NITD-349 is an MmpL3 inhibitor that shows highly potent anti-mycobacterial activity with MIC50 of 23 nM against virulent Mycobacterium tuberculosis H37Rv. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.