Learn More
Medchemexpress LLC NITD-688 | 2407227-31-8 | 98.0% | 500.68 | 100 MG
SDP
Catalog No. 5000350140
Encompass_Preferred
NITD-688 is an orally active pan-serotype inhibitor of the dengue virus NS4B protein and can be used in the research of dengue virus (DENV).
  • Orally active pan-serotype inhibitor of the dengue virus NS4B protein.
  • Exhibits antiviral activity against all four serotypes of dengue virus (DENV), with EC50 values ranging from 8 to 38 nM.
  • Shows antiviral activity against DENV-2 infected PBMCs, with an EC50 value of 0.94 nM.
  • Directly binds to wild-type NS4B protein.
  • Demonstrates excellent antiviral efficacy in infected AG129 mice.
Catalog No. 50-003-50140 Supplier Medchemexpress LLC Supplier No. HY138543100MG
NITD-688 is an orally active pan-serotype inhibitor of the dengue virus NS4B protein and can be used in the research of dengue virus (DENV).
  • Orally active pan-serotype inhibitor of the dengue virus NS4B protein.
  • Exhibits antiviral activity against all four serotypes of dengue virus (DENV), with EC50 values ranging from 8 to 38 nM.
  • Shows antiviral activity against DENV-2 infected PBMCs, with an EC50 value of 0.94 nM.
  • Directly binds to wild-type NS4B protein.
  • Demonstrates excellent antiviral efficacy in infected AG129 mice.