Learn More
Medchemexpress LLC NITD-688 | 2407227-31-8 | 98.0% | 500.68 | 25 MG
SDP
Catalog No. 5000348613
Encompass_Preferred
NITD-688 is an orally active pan-serotype inhibitor of the dengue virus NS4B protein. It is utilized in dengue virus (DENV) research and demonstrates antiviral activity against all four serotypes of DENV.
  • Orally active pan-serotype inhibitor of the dengue virus NS4B protein
  • Utilized in dengue virus (DENV) research
  • Exhibits antiviral activity against all four serotypes of DENV
  • Shows antiviral activity against DENV-2 infected PBMCs with an EC50 value of 0.94 nM
  • Directly binds to wild-type but not mutant NS4B protein
  • Demonstrates antiviral efficacy in infected AG129 mice
  • Well tolerated in rats and dogs
Catalog No. 50-003-48613 Supplier Medchemexpress LLC Supplier No. 25MGHY138543
NITD-688 is an orally active pan-serotype inhibitor of the dengue virus NS4B protein. It is utilized in dengue virus (DENV) research and demonstrates antiviral activity against all four serotypes of DENV.
  • Orally active pan-serotype inhibitor of the dengue virus NS4B protein
  • Utilized in dengue virus (DENV) research
  • Exhibits antiviral activity against all four serotypes of DENV
  • Shows antiviral activity against DENV-2 infected PBMCs with an EC50 value of 0.94 nM
  • Directly binds to wild-type but not mutant NS4B protein
  • Demonstrates antiviral efficacy in infected AG129 mice
  • Well tolerated in rats and dogs