Learn More
Medchemexpress LLC Nitd-688 | 2407227-31-8 | 98.0% | 500.68 g·mol⁻¹ | C25H32N4O3S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1385435MG
Encompass_Preferred
NITD-688 is an orally active small-molecule pan-serotype inhibitor of the dengue virus NS4B protein for research use. It exhibits potent activity against all four dengue virus serotypes and has demonstrated favorable pharmacokinetics and in vivo efficacy in preclinical studies. The compound is supplied as a white to off-white solid with reported high purity and recommended cold-storage conditions for stability.
  • Pan-serotype NS4B inhibition
  • Potent antiviral activity against all four DENV serotypes
  • Demonstrated oral efficacy in preclinical models
  • High reported purity (≈98%)
  • Supplied as a solid with cold-storage stability
Catalog No. 50-002-22970