Learn More
Medchemexpress LLC NITD-688 | 2407227-31-8 | 98.0% | C25H32N4O3S2 | 10MG
SDP
Catalog No. 5000260582
Encompass_Preferred
NITD-688 is a small-molecule, orally active pan-serotype inhibitor of the dengue virus NS4B protein intended for preclinical and research use. It shows low-nanomolar antiviral potency across all four DENV serotypes and has demonstrated oral efficacy in infected AG129 mice, making it suitable for antiviral profiling and mechanistic studies of NS4B biology.
  • Potent pan-serotype DENV inhibitor with low-nanomolar activity.
  • Orally active in preclinical efficacy studies.
  • Directly binds NS4B to inhibit viral replication.
  • Supplied as a white to off-white solid for research use.
  • High purity supporting reproducible experimental results.
  • Useful for antiviral profiling and mechanistic NS4B studies.
Catalog No. 50-002-60582 Supplier Medchemexpress LLC Supplier No. HY13854310MG
NITD-688 is a small-molecule, orally active pan-serotype inhibitor of the dengue virus NS4B protein intended for preclinical and research use. It shows low-nanomolar antiviral potency across all four DENV serotypes and has demonstrated oral efficacy in infected AG129 mice, making it suitable for antiviral profiling and mechanistic studies of NS4B biology.
  • Potent pan-serotype DENV inhibitor with low-nanomolar activity.
  • Orally active in preclinical efficacy studies.
  • Directly binds NS4B to inhibit viral replication.
  • Supplied as a white to off-white solid for research use.
  • High purity supporting reproducible experimental results.
  • Useful for antiviral profiling and mechanistic NS4B studies.