Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NITD-916 | 1614262-83-7 | 99.8% | 1 ML
SDP

Catalog No. 5000230134
Encompass_Preferred

NITD-916 is a 4-hydroxy-2-pyridone derivative that inhibits the mycobacterial enoyl reductase InhA (IC50 570 nM) and demonstrates potent anti-tuberculosis activity. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for research use and should be stored and handled according to the SDS.

  • High purity, 99.83%.
  • Molecular formula C20H25NO2; molecular weight 311.42 g/mol.
  • Potent InhA inhibition (IC50 570 nM) and cellular activity (MIC50 ~50 nM).
  • Provided as a 10 mM solution in DMSO for convenient dosing.
  • Store powder at -20°C; store in solvent at -80°C for long-term stability.

Catalog No. 50-002-30134 Supplier Medchemexpress LLC Supplier No. HY12264310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NITD-916 is a 4-hydroxy-2-pyridone derivative that inhibits the mycobacterial enoyl reductase InhA (IC50 570 nM) and demonstrates potent anti-tuberculosis activity. Supplied as a 10 mM solution in DMSO (1 mL), it is intended for research use and should be stored and handled according to the SDS.

  • High purity, 99.83%.
  • Molecular formula C20H25NO2; molecular weight 311.42 g/mol.
  • Potent InhA inhibition (IC50 570 nM) and cellular activity (MIC50 ~50 nM).
  • Provided as a 10 mM solution in DMSO for convenient dosing.
  • Store powder at -20°C; store in solvent at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.