Learn More
TARGETMOL CHEMICALS INC NITD-916 5MG
SDP
Catalog No. 502579587
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. NITD-916 is an orally active and highly lipophilic inhibitor of Mycobacterium enoyl reductase InhA with an IC50 of 570 nM.NITD-916 is a potent 4-hydroxy-2-pyridone derivative that forms a ternary complex with InhA and NADH preventing access to the fatty acyl substrate-binding pocket.NITD-916 exhibits antituberculosis activity. purity: 99%
Catalog No. 50-257-9587 Supplier TARGETMOL CHEMICALS INC Supplier No. T607745MG
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. NITD-916 is an orally active and highly lipophilic inhibitor of Mycobacterium enoyl reductase InhA with an IC50 of 570 nM.NITD-916 is a potent 4-hydroxy-2-pyridone derivative that forms a ternary complex with InhA and NADH preventing access to the fatty acyl substrate-binding pocket.NITD-916 exhibits antituberculosis activity. purity: 99%