Learn More
Carson Optical NiteMax Digital Night Vision Monocular
SDPSCI_ED
Catalog No. S001480 Shop All Carson Optical Products
Change view
Click to view available options
Magnification:
1.8x
Catalog No. Magnification
S001480 1.8x
1 options
Catalog No. S001480 Supplier Carson Optical Supplier No. NV300
Add to Cart
Add to Cart
The NiteMax Digital Night Vision Monocular uses infrared illumination for low-light viewing.
  • Digital night vision monocular with 1.8x magnification for low-light conditions and total darkness
  • Built-in infrared illuminator with three adjustable intensity settings
  • 1.5 in. (38 mm) viewing screen
  • Manual focusing dial
  • Easy-to-use controls for beginners
  • Automatic power saving: screen dims after approximately 2 minutes and the unit shuts off after approximately 2 minutes 30 seconds
  • Operates on three AAA batteries (not included)
  • Compact, lightweight body fits in a pocket, backpack or vehicle
  • Suitable for wildlife observation, camping, nature walks and nighttime property observation
Specifications
Magnification 1.8x