Learn More
Medchemexpress LLC HY-B0820 100mg Medchemexpress, Nitenpyram CAS:150824-47-8 Purity:>98%
SDP
Catalog No. 501970126
Encompass_Preferred
Medchemexpress, HY-B0820 100mg Nitenpyram CAS:150824-47-8 Nitenpyram is a calss of neonicotinoid and an insect nicotinic acetylcholine receptor (nAChR) agonist with an IC50 of 14 nM. Nitenpyram is an oral fast-acting insecticide used to suppress sucking insects on companion animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0126 Supplier Medchemexpress LLC Supplier No. HYB0820100MG
Medchemexpress, HY-B0820 100mg Nitenpyram CAS:150824-47-8 Nitenpyram is a calss of neonicotinoid and an insect nicotinic acetylcholine receptor (nAChR) agonist with an IC50 of 14 nM. Nitenpyram is an oral fast-acting insecticide used to suppress sucking insects on companion animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.