Learn More
Cayman Chemical NItenpyram-d3 500ug
SDP
Catalog No. 502932942
Encompass_Preferred
An internal standard for the quantification of nitenpyram by GC- or LC-MS
Catalog No. 50-293-2942 Supplier Cayman Chemical Supplier No. 26783500 UG
An internal standard for the quantification of nitenpyram by GC- or LC-MS