Learn More
Wildlife Supply Company Wildco Bolt Cloth-Nitex 100 μm 40" W
SDP
Catalog No. NC1582054
Encompass_Preferred
The world standard for plankton nets due to its consistency and ability to maintain aperture size and shape. Precision woven from high quality, durable nylon threads. Both warp and weft thread diameters maintain square aperture dimensions. After weaving, this cloth is “calendared” - run between hot rollers to heat set the woven threads in their exact positions. Nitex with an aperture of 600 microns or more is stiffer.
Catalog No. NC1582054 Supplier Wildlife Supply Company Wildco Supplier No. 324C34
The world standard for plankton nets due to its consistency and ability to maintain aperture size and shape. Precision woven from high quality, durable nylon threads. Both warp and weft thread diameters maintain square aperture dimensions. After weaving, this cloth is “calendared” - run between hot rollers to heat set the woven threads in their exact positions. Nitex with an aperture of 600 microns or more is stiffer.