Learn More
Genesee Scientific Corporation NITEX NYLON MESH 630 MICRON
SDP
Catalog No. NC9432449
Encompass
Mesh, Nitex Nylon 630um 45" Roll 1 Foot/Unit Ideal for catching adults when collecting embryos, this Nitex nylon mesh has a 630�m pore size and 53% open area. Sold by the contiguous lineal foot from a 45" roll. From Genesee Scientific's extensive line of Drosophila products, this mesh is sold by the lineal foot, so you can order just as much as you need.
Catalog No. NC9432449 Supplier Genesee Scientific Corporation Supplier No. 57101
Mesh, Nitex Nylon 630um 45" Roll 1 Foot/Unit Ideal for catching adults when collecting embryos, this Nitex nylon mesh has a 630�m pore size and 53% open area. Sold by the contiguous lineal foot from a 45" roll. From Genesee Scientific's extensive line of Drosophila products, this mesh is sold by the lineal foot, so you can order just as much as you need.