Learn More
Apexbio Technology LLC NITIDINE-CHLORIDE-20MG
SDP
Catalog No. 5000570713
Encompass_Preferred
Nitidine chloride (CAS No 13063-04-2) is a bioactive alkaloid derived from the roots of Zanthoxylum species It is extensively researched within the pharmacological and biomedical fields due to its diverse biological activities The compound primarily exerts its effects by inhibiting pathways such as topoisomerases and interacting with DNA contributing to its antitumor and anti-inflammatory properties In vitro it demonstrates efficacy in the nanomolar to low micromolar range against cancer cell lines Nitidine chloride is frequently employed in cancer research utilizing cell models and animal studies to evaluate its therapeutic potential It is valuable in drug screening for novel anticancer agents Experimental concentrations typically depend on the specific research design emphasizing the compound s adaptability to various investigative scenarios
Catalog No. 50-005-70713 Supplier Apexbio Technology LLC Supplier No. N262620
Nitidine chloride (CAS No 13063-04-2) is a bioactive alkaloid derived from the roots of Zanthoxylum species It is extensively researched within the pharmacological and biomedical fields due to its diverse biological activities The compound primarily exerts its effects by inhibiting pathways such as topoisomerases and interacting with DNA contributing to its antitumor and anti-inflammatory properties In vitro it demonstrates efficacy in the nanomolar to low micromolar range against cancer cell lines Nitidine chloride is frequently employed in cancer research utilizing cell models and animal studies to evaluate its therapeutic potential It is valuable in drug screening for novel anticancer agents Experimental concentrations typically depend on the specific research design emphasizing the compound s adaptability to various investigative scenarios