Learn More
Educational Innovations Nitinol Spring
SDP
Catalog No. NC1376032
Encompass
What's so special about a piece of coiled wire? Plenty. Our spring is made of nitinol, an amazing metal that can be twisted or straightened but will always return to its original shape when dipped into hot water (~50°C). Talk about discrepant events - this wire is practically magic! An alloy of nickel and titanium, nitinol (Ni-Ti) falls into a sophisticated class of materials known as shape memory alloys that "remember" their original shape. Perfect for classroom demonstrations. Comes with a Teacher's Guide. Wire is 0.75 mm thick, with a 20 mm inner diameter and 45 cm overall length
Catalog No. NC1376032 Supplier Educational Innovations Supplier No. HS620
What's so special about a piece of coiled wire? Plenty. Our spring is made of nitinol, an amazing metal that can be twisted or straightened but will always return to its original shape when dipped into hot water (~50°C). Talk about discrepant events - this wire is practically magic! An alloy of nickel and titanium, nitinol (Ni-Ti) falls into a sophisticated class of materials known as shape memory alloys that "remember" their original shape. Perfect for classroom demonstrations. Comes with a Teacher's Guide. Wire is 0.75 mm thick, with a 20 mm inner diameter and 45 cm overall length