Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitinol wire, 1.0mm (0.039 in.) dia., shape memory, Alloy M, straight annealed, oxide surface, Thermo Scientific™

Catalog No. AA45658G6
Change view
Click to view available options
Quantity:
10 m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45658G6 10 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45658G6 Supplier Thermo Scientific Chemicals Supplier No. 45658G6

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitinol wire, Alloy M
Name Note 1.0mm (0.039 in.) dia., Shape memory, Straight annealed, Oxide surface
Quantity 10 m
Odor Odorless
Weight ≈49.71g/10m
MDL Number MFCD02091734
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.