Learn More
TARGETMOL CHEMICALS INC Nitisinone 100MG
SDP
Catalog No. 502877399
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Nitisinone (SC0735)(SC0735) is an enzyme 4-hydroxyphenylpyruvate dioxygenase inhibitor. Purity 99.54%
Catalog No. 50-287-7399 Supplier TARGETMOL CHEMICALS INC Supplier No. T1684100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Nitisinone (SC0735)(SC0735) is an enzyme 4-hydroxyphenylpyruvate dioxygenase inhibitor. Purity 99.54%