Learn More
Medchemexpress LLC Nitisinone | 104206-65-7 | 100 MG
SDP
Catalog No. 5000408383
Encompass_Preferred
Nitisinone is an orally active, competitive, and reversible 4-hydroxyphenylpyruvate dioxygenase (4-HPPD) inhibitor with an IC50 of 173 nM. It promotes tyrosine accumulation in a dose-dependent manner and can be used in studies of hereditary tyrosinemia type 1 (HT-1) and albinism.
  • Orally active and competitive 4-HPPD inhibitor
  • Promotes tyrosine accumulation
  • Useful in studies of hereditary tyrosinemia type 1 (HT-1)
  • Applicable in studies of albinism
Catalog No. 50-004-08383 Supplier Medchemexpress LLC Supplier No. HYB0607100MG
Nitisinone is an orally active, competitive, and reversible 4-hydroxyphenylpyruvate dioxygenase (4-HPPD) inhibitor with an IC50 of 173 nM. It promotes tyrosine accumulation in a dose-dependent manner and can be used in studies of hereditary tyrosinemia type 1 (HT-1) and albinism.
  • Orally active and competitive 4-HPPD inhibitor
  • Promotes tyrosine accumulation
  • Useful in studies of hereditary tyrosinemia type 1 (HT-1)
  • Applicable in studies of albinism