Learn More
Apexbio Technology LLC Nitisinone 104206-65-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089658
Encompass_Preferred
Nitisinone (CAS 104206-65-7) is a synthetic small molecule that selectively inhibits 4-hydroxyphenylpyruvate dioxygenase an enzyme involved in the catabolic pathway of tyrosine By blocking this enzyme nitisinone disrupts the conversion of 4-hydroxyphenylpyruvate to homogentisic acid leading to reduced production of toxic tyrosine metabolites This mechanism is utilized in studies of hereditary tyrosinemia type 1 where nitisinone serves as a pharmacological tool to investigate metabolic regulation and therapeutic intervention in tyrosine degradation disorders
Catalog No. 50-000-89658 Supplier Apexbio Technology LLC Supplier No. B35275.1
Nitisinone (CAS 104206-65-7) is a synthetic small molecule that selectively inhibits 4-hydroxyphenylpyruvate dioxygenase an enzyme involved in the catabolic pathway of tyrosine By blocking this enzyme nitisinone disrupts the conversion of 4-hydroxyphenylpyruvate to homogentisic acid leading to reduced production of toxic tyrosine metabolites This mechanism is utilized in studies of hereditary tyrosinemia type 1 where nitisinone serves as a pharmacological tool to investigate metabolic regulation and therapeutic intervention in tyrosine degradation disorders