Learn More
Medchemexpress LLC Nitisinone | 104206-65-7 | 99.93% | 1 ML
SDP
Catalog No. 5000407774
Encompass_Preferred
Nitisinone is an orally active, competitive, and reversible 4-hydroxyphenylpyruvate dioxygenase (4-HPPD) inhibitor with an IC50 of 173 nM. It promotes tyrosine accumulation in a dose-dependent manner.
  • Used in studies of hereditary tyrosinemia type 1 (HT-1)
  • Used in studies of albinism
  • Orally active
  • Competitive and reversible 4-hydroxyphenylpyruvate dioxygenase (4-HPPD) inhibitor
Catalog No. 50-004-07774 Supplier Medchemexpress LLC Supplier No. HYB060710MM/1ML
Nitisinone is an orally active, competitive, and reversible 4-hydroxyphenylpyruvate dioxygenase (4-HPPD) inhibitor with an IC50 of 173 nM. It promotes tyrosine accumulation in a dose-dependent manner.
  • Used in studies of hereditary tyrosinemia type 1 (HT-1)
  • Used in studies of albinism
  • Orally active
  • Competitive and reversible 4-hydroxyphenylpyruvate dioxygenase (4-HPPD) inhibitor