Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Nitrate Broth Prepared Media with Durham Tube
SDPSCI_ED

Catalog No. S42833 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S42833 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S42833 Supplier Carolina™ Supplier No. 826082

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Carefully tested for sterility and ability to support the characteristic growth of appropriate organisms.

  • Sterile
  • Ready-to-use media
  • Used for characteristic growth of organisms

Specifications

Content And Storage 2°C to 8°C
Description Sterile
Manufacturing Origin US with Globally Sourced Materials
Media Type Nitrate broth with prepared media
Product Type Nutrient Broth
Sterility Sterile
Target Organism Class Fastidious and Non-fastidious Organisms
Test Type Microbial Cultivation
For Use With (Application) Culture of organisms
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.