Learn More
Hach Company Nitrate Ionic Strength Adjustor (ISA) Buffer Solution, 500 mL
Supplier:  Hach Company 2488349
Encompass
For sample treatment in nitrate determination by ion selective electrode. 500 mL bottle.
Catalog No. 50-199-6058