Learn More
Hach Company Nitrate Ionic Strength Adjustor (ISA) Powder Pillows, pk/100
Catalog No. 501997039 Shop All Hach Company Products
Encompass
Ionic Strength Adjustor (ISA) for Nitrate determinations by Ion Selective Electrode (ISE) method. Powder form prevent from diluting samples. Ideal for low Nitrate level readings. Eliminate most interferences. Pack of 100 powder pillows.
Catalog No. 50-199-7039 Supplier Hach Company Supplier No. 4456369
Ionic Strength Adjustor (ISA) for Nitrate determinations by Ion Selective Electrode (ISE) method. Powder form prevent from diluting samples. Ideal for low Nitrate level readings. Eliminate most interferences. Pack of 100 powder pillows.