Learn More
Hach Company Nitrate Ionic Strength Adjustor Powder Pillows - pk/100
Catalog No. 501996998 Shop All Hach Company Products
Encompass
Ionic Strength Adjustor for use with IntelliCAL ISENO3181 or sensION+ 9663 nitrate ion selective electrodes. The ISA is critical for determining low level nitrate concentrations. Powder form does not dilute samples. Pack of 100 Powder Pillows.
Catalog No. 50-199-6998 Supplier Hach Company Supplier No. 2984799
Ionic Strength Adjustor for use with IntelliCAL ISENO3181 or sensION+ 9663 nitrate ion selective electrodes. The ISA is critical for determining low level nitrate concentrations. Powder form does not dilute samples. Pack of 100 Powder Pillows.