Learn More
AdipoGen Nitrate Ionophore VI
SDP
Catalog No. 502560773
Encompass_Preferred
Chemical. CAS 1196157-85-3. Formula C28H60N4O4. MW 516.8. Synthetic. Ionophore for online monitoring of nitrate with superior characteristics. Detection limit <200µg NO3-/L. Ion selectivity logKpotNO3/NO2-1.2, logKpotNO3/Cl-2.5, logKpotNO3/Br-1.1, logKpotNO3/HCO3-3.2, logKpotNO3/HPO4-4.4, logKpotNO3/SO4-4.5
Catalog No. 50-256-0773 Supplier AdipoGen Supplier No. CDXH0054M010
Chemical. CAS 1196157-85-3. Formula C28H60N4O4. MW 516.8. Synthetic. Ionophore for online monitoring of nitrate with superior characteristics. Detection limit <200µg NO3-/L. Ion selectivity logKpotNO3/NO2-1.2, logKpotNO3/Cl-2.5, logKpotNO3/Br-1.1, logKpotNO3/HCO3-3.2, logKpotNO3/HPO4-4.4, logKpotNO3/SO4-4.5