Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Nitrate-Nitrogen Tablet Test Kit
SDPSCI_ED

Catalog No. S65410 Shop All LaMotte Company Products
Change view
Click to view available options
Range:
1 to 15 ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S65410 1 to 15 ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65410 Supplier LaMotte™ Supplier No. 335401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

50 Nitrate No. 1 tablets, 50 Nitrate No. 2 test tablets, two plastic test tubes with caps, Octa-Slide viewer with slide bar, instructions

The Nitrate-Nitrogen Tablet Test Kit is a colorimetric test system.

  • Range: 0 to 15 ppm
  • Sensitivity: 1.0 ppm
  • Number of Tests: 50
  • Reagent Refills for use with Nitrate-Nitrogen Kit (3354-01)

Specifications

For Use With (Application) Drinking water, environmental education, food and beverage, industrial water boiling and cooling, laundry and sanitation, water and wastewater
Includes 50 nitrate No. 1 tablets, 50 nitrate No. 2 test tablets, two plastic test tubes with caps, Octa slide viewer with slide bar, instructions
No. of Tests 50
Range 1 to 15 ppm
Test Method Zinc Reduction
Type Nitrate Nitrogen Tablet Test Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.