Learn More
Hach Company Intellical™ ISENO3181 Nitrate (NO₃⁻) Ion Selective Electrode (ISE), 1 m cable
Catalog No. 501997733 Shop All Hach Company Products
Encompass
Intellical™ ISENO3181 is a digital, combination, Nitrate ion selective electrode (ISE) with a with a non-refillable gel Dritek reference with a double junction, ceramic porous pin and porous PTFE annular ring, and built-in temperature sensor. The electrode measures Nitrate concentration in water samples. The laboratory version of this ISE is shockproff with its Epoxy plastic body.The Intellical ISENO3181 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISENO3181’s solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE. The ISENO3181 is ideal for measuring nitrate concentrations in wastewater, drinking water and general water quality applications.
Catalog No. 50-199-7733 Supplier Hach Company Supplier No. ISENO318101
Intellical™ ISENO3181 is a digital, combination, Nitrate ion selective electrode (ISE) with a with a non-refillable gel Dritek reference with a double junction, ceramic porous pin and porous PTFE annular ring, and built-in temperature sensor. The electrode measures Nitrate concentration in water samples. The laboratory version of this ISE is shockproff with its Epoxy plastic body.The Intellical ISENO3181 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISENO3181’s solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE. The ISENO3181 is ideal for measuring nitrate concentrations in wastewater, drinking water and general water quality applications.