Learn More
Nitrate, Quant™ Test Strips
Catalog No. AA23282QA
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
AA23282QA 1 Kit
1 options
Catalog No. AA23282QA Supplier Thermo Scientific Chemicals Supplier No. 023282QA
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
What should I do if the reaction zone color on the Nitrate, Quant Test Strip is the same as or darker than the darkest color of the color scale?
If the color of the reaction zone is the same as or darker than the darkest color of the scale, we recommend that you repeat the test using fresh, diluted sample until a result of less than 500 mg/L nitrate ion is observed.
What protocol should I follow when using the Nitrate, Quant Test Strips?
We recommend the following protocol to use the Nitrate, Quant Test Strips:
1. Take one strip from the container and immediately close the container.
2. Immerse the reaction zones of the test strip in the pretreated sample for 1 second. The sample should be 59-77°F (15 - 25°C).
3. Remove the test strip and shake off excess liquid from the strip.
4. Wait 1 minute and then compare the color field on the label with the reaction zone on the Nitrate test strip.
Note: Samples containing more than 500 mg/L nitrate ion must be diluted with distilled water. The pH must be between 1-12. If the pH is lower than 1, add sodium acetate. If the pH is greater than 12, adjust with tartaric acid. Extract solid samples by an appropriate method.
How should my sample be prepared for use with the Nitrate, Quant Test Strips?
Samples containing more than 500 mg/L nitrate ion must be diluted with distilled water. The pH must be between 1-12. If the pH is lower than 1, add sodium acetate. If the pH is greater than 12, adjust with tartaric acid. Extract solid samples by an appropriate method.
How many test strips come in a pack of Nitrate, Quant Test Strips?
100 test strips come in a pack of Nitrate, Quant Test Strips.
What are the graduations for the Nitrate, Quant Test Strips?
The Nitrate, Quant Test Strips have the following graduations: 0-10-25-50-100-250-500 mg/L nitrate ion.
When should I use the Nitrate, Quant Test Strips?
The Nitrate, Quant Test Strips are designed for preliminary tests to determine the graduated concentration ranges of nitrate ion.