Learn More
Alkali Scientific Nitrate Reagent B, 25 mL (1 oz), Amber Bottle, 1/Pk
SDP
Catalog No. 5000663061
Encompass_Preferred
Nitrate Reagent B, 25 mL (1 oz), Amber Bottle, 1/Pk. Nitrate Reagent B is used in conjunction with Nitrate Reagent A for nitrate reduction testing. It helps differentiate bacterial species based on their ability to reduce nitrates to nitrites. The amber bottle ensures the reagent remains protected from light, maintaining its efficacy and reliability for laboratory use.
Catalog No. 50-006-63061 Supplier Alkali Scientific Supplier No. NRAB00
Nitrate Reagent B, 25 mL (1 oz), Amber Bottle, 1/Pk. Nitrate Reagent B is used in conjunction with Nitrate Reagent A for nitrate reduction testing. It helps differentiate bacterial species based on their ability to reduce nitrates to nitrites. The amber bottle ensures the reagent remains protected from light, maintaining its efficacy and reliability for laboratory use.