Learn More
CHEMetrics™ by AquaPhoenix Nitrate Refill Kit

Description
- Nitrate is reduced to nitrite by shaking the sample with a zinc metal catalyst and a citrate buffer.
- The nitrite is reacted with sulfanilic acid and N-(1-naphthyl)ethylenediamine dihydrochloride (NED) in an acidic solution to produce a intense reddish-purple azo dye. Results are expressed as ppm (mg/L) nitrate-nitrogen (NO3-N). This method is applicable to industrial wastewaters, drinking, and surface waters. These Nitrate in water test kits can also be used for the analysis of seawater. To convert results from ppm NO3-N to ppm NO3, multiply by 4.43.
Specifications
Specifications
| pH Range | 7 |
| Volume (Metric) | 0.5 mL |
| For Use With (Equipment) | K-6901, K-6901A or K-6901D |
| No. of Tests | 30 |
| Quantity | 30/Box |
| Type | Nitrate Refill Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.