Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrate Standard, 1000 ppm NO3 (226 ppm N), Reagents

Catalog No. 01334659
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-659 120 mL
01-334-658 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-659 Supplier Reagents Holdings Llc Supplier No. CN103100120A
 Restricted Item
Add to Cart
Add to Cart

Nitrate Standard, 1000 ppm NO3 (226 ppm N), Reagent, use for Nitrogen analysis. Manufactured in ISO 9001 facility.

Specifications

Quantity 120 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.