Learn More
SPEX CERTIPREP LLC 3P N-NO3 CRM, 125 mL
Catalog No. NC1470779
Encompass_Preferred
(NSI by Zeptometrix) 1000mg/L in Water. Derived from high purity Sodium nitrate, NIST traceable.
Catalog No. NC1470779 Supplier SPEX CERTIPREP LLC 3P Supplier No. IS011
(NSI by Zeptometrix) 1000mg/L in Water. Derived from high purity Sodium nitrate, NIST traceable.