Learn More
Hach Company Hach Nitrate Standard 25 mg/L NO3 (5.65 mg/L NO3-N), 500 mL (LCW828)
Catalog No. NC2793522 Shop All Hach Company Products
Encompass
Hach nitrate standard solution at 25 mg/L NO3 (5.65 mg/L NO3-N) supplied in a 500 mL bottle for calibration and verification of online nitrate monitoring equipment and sensors.
  • Concentration: 25 mg/L NO3 (5.65 mg/L NO3-N).
  • Volume: 500 mL.
  • Intended use: standard solution for online nitrate determination and instrument calibration.
  • Certificates of analysis are available for download.
  • Safety data sheets are available; follow standard laboratory handling and storage procedures.
Catalog No. NC2793522 Supplier Hach Company Supplier No. LCW828
Hach nitrate standard solution at 25 mg/L NO3 (5.65 mg/L NO3-N) supplied in a 500 mL bottle for calibration and verification of online nitrate monitoring equipment and sensors.
  • Concentration: 25 mg/L NO3 (5.65 mg/L NO3-N).
  • Volume: 500 mL.
  • Intended use: standard solution for online nitrate determination and instrument calibration.
  • Certificates of analysis are available for download.
  • Safety data sheets are available; follow standard laboratory handling and storage procedures.