Learn More
SPEX CERTIPREP LLC 3P NITRATE STD 1000PPM 250ML
Catalog No. NC0211467
Encompass_Preferred
(HPS by Zeptometrix) 1000 ug/mL Nitrate from NaNO3, 99.999% (metals basis), in H2O; 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohn water, packaged in HDPE bottles.
Catalog No. NC0211467 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICNOM250
(HPS by Zeptometrix) 1000 ug/mL Nitrate from NaNO3, 99.999% (metals basis), in H2O; 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohn water, packaged in HDPE bottles.