Learn More
Aqua Solutions Nitrate Standard 1,000 ppm as N0₃ (1ml = 1mg NO₃) (100ML)
SDP
Catalog No. NC9194954
Encompass_Preferred
Nitrate Standard 1,000 ppm as N0₃ (1ml = 1mg NO₃) (100ML)
Catalog No. NC9194954 Supplier Aqua Solutions Supplier No. 6478100ML
Nitrate Standard 1,000 ppm as N0₃ (1ml = 1mg NO₃) (100ML)